siRNA Details
| Gene Symbol: Vom1r67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008912.1 | NP_001008912.1 | 1 | 595 | ||
| GenBank mRNA | AY510295.1 | - | 1 | 595 | ||
| Gene Symbol: Vom1r66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008945.1 | NP_001008945.1 | 1 | 595 | ||
| GenBank mRNA | AY510310.1 | - | 1 | 595 | ||

Vom1r67
Genomic Map