siRNA Details
| Gene Symbol: Vom1r36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008913.1 | NP_001008913.1 | 1 | 180 | ||
| GenBank mRNA | AY510335.1 | - | 1 | 180 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom1r36| Gene Symbol: Vom1r36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008913.1 | NP_001008913.1 | 1 | 180 | ||
| GenBank mRNA | AY510335.1 | - | 1 | 180 | ||