siRNA Details
| Gene Symbol: Vom1r47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008916.1 | NP_001008916.1 | 1 | 607 | ||
| GenBank mRNA | AY510316.1 | - | 1 | 607 | ||
| AY510317.1 | - | 1 | 607 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom1r47| Gene Symbol: Vom1r47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008916.1 | NP_001008916.1 | 1 | 607 | ||
| GenBank mRNA | AY510316.1 | - | 1 | 607 | ||
| AY510317.1 | - | 1 | 607 | |||