siRNA Details
| Gene Symbol: Vom1r49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008920.1 | NP_001008920.1 | 550 | |||
| GenBank mRNA | AY510303.1 | - | 550 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom1r49| Gene Symbol: Vom1r49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008920.1 | NP_001008920.1 | 550 | |||
| GenBank mRNA | AY510303.1 | - | 550 | |||