siRNA Details
| Gene Symbol: Vom1r23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008941.1 | NP_001008941.1 | 1 | 477 | ||
| GenBank mRNA | AY510359.1 | - | 1 | 477 | ||
| Gene Symbol: Vom1r-ps112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758822.1 | XP_008757044.1 | 1 | 539 | ||

Vom1r23
Genomic Map