siRNA Details
| Gene Symbol: Vom1r95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008950.1 | NP_001008950.1 | 1 | 776 | ||
| GenBank mRNA | AY510341.1 | - | 1 | 776 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom1r95| Gene Symbol: Vom1r95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008950.1 | NP_001008950.1 | 1 | 776 | ||
| GenBank mRNA | AY510341.1 | - | 1 | 776 | ||