siRNA Details
| Gene Symbol: Vom1r10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008958.1 | NP_001008958.1 | 1 | 668 | ||
| GenBank mRNA | AY510358.1 | - | 1 | 668 | ||
| Gene Symbol: Vom1r14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008961.1 | NP_001008961.1 | 1 | 668 | ||
| GenBank mRNA | AY510357.1 | - | 1 | 668 | ||

Vom1r10
Genomic Map