siRNA Details
| Gene Symbol: Vom1r28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008966.1 | NP_001008966.1 | 1 | 524 | ||
| GenBank mRNA | AY510353.1 | - | 1 | 524 | ||
| Gene Symbol: LOC103691019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758872.1 | XP_008757094.1 | 1 | 524 | ||

Vom1r28
Genomic Map