siRNA Details
| Gene Symbol: Vom1r92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008968.1 | NP_001008968.1 | 1 | 205 | ||
| XM_006236847.1 | XP_006236909.1 | 2 | 280 | |||
| GenBank mRNA | AY510279.1 | - | 1 | 205 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom1r92| Gene Symbol: Vom1r92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008968.1 | NP_001008968.1 | 1 | 205 | ||
| XM_006236847.1 | XP_006236909.1 | 2 | 280 | |||
| GenBank mRNA | AY510279.1 | - | 1 | 205 | ||