siRNA Details
| Gene Symbol: Vom1r31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008969.1 | NP_001008969.1 | 613 | |||
| GenBank mRNA | AY510343.1 | - | 613 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom1r31| Gene Symbol: Vom1r31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008969.1 | NP_001008969.1 | 613 | |||
| GenBank mRNA | AY510343.1 | - | 613 | |||