siRNA Details
| Gene Symbol: Vom1r75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008972.1 | NP_001008972.1 | 1 | 208 | ||
| GenBank mRNA | AY510283.1 | - | 1 | 208 | ||
| Gene Symbol: LOC100909473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006236570.1 | XP_006236632.1 | 1 | 208 | ||

Vom1r75
Genomic Map