siRNA Details
| Gene Symbol: Ccdc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009533.2 | NP_001009533.2 | 1 | 893 | ||
| GenBank mRNA | BC060520.1 | - | 1 | 893 | 5597839 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc8| Gene Symbol: Ccdc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009533.2 | NP_001009533.2 | 1 | 893 | ||
| GenBank mRNA | BC060520.1 | - | 1 | 893 | 5597839 | |