siRNA Details
| Gene Symbol: Pms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009535.1 | NP_001009535.1 | 7 | 869 | ||
| GenBank mRNA | BC061722.1 | - | 7 | 869 | 5598508 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pms1| Gene Symbol: Pms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009535.1 | NP_001009535.1 | 7 | 869 | ||
| GenBank mRNA | BC061722.1 | - | 7 | 869 | 5598508 | |