siRNA Details
| Gene Symbol: Zfp799 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241084.2 | XP_006241146.1 | 4 | 1928 | ||
| GenBank mRNA | BC061577.1 | - | 4 | 1932 | 6888485 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp799| Gene Symbol: Zfp799 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241084.2 | XP_006241146.1 | 4 | 1928 | ||
| GenBank mRNA | BC061577.1 | - | 4 | 1932 | 6888485 | |