siRNA Details
| Gene Symbol: Ier2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009541.1 | NP_001009541.1 | 1 | 1476 | ||
| GenBank mRNA | BC061717.1 | - | 1 | 1476 | 5598375 | |
| BC158596.1 | - | 2 | 1421 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ier2| Gene Symbol: Ier2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009541.1 | NP_001009541.1 | 1 | 1476 | ||
| GenBank mRNA | BC061717.1 | - | 1 | 1476 | 5598375 | |
| BC158596.1 | - | 2 | 1421 | |||