siRNA Details
| Gene Symbol: Slmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009543.1 | NP_001009543.1 | 3 | 474 | ||
| GenBank mRNA | BC060590.1 | - | 3 | 474 | 6918453 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slmo2| Gene Symbol: Slmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009543.1 | NP_001009543.1 | 3 | 474 | ||
| GenBank mRNA | BC060590.1 | - | 3 | 474 | 6918453 | |