siRNA Details
| Gene Symbol: Srsf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009720.2 | NP_001009720.1 | 2 | 670 | ||
| XM_006247813.2 | XP_006247875.1 | 2 | 697 | |||
| XM_008768382.1 | XP_008766604.1 | 2 | 697 | |||
| GenBank mRNA | BC058508.1 | - | 2 | 674 | 6889746 | |
| FQ232539.1 | - | 2 | 671 | |||

Srsf2
Genomic Map