siRNA Details
| Gene Symbol: Ggnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009972.1 | NP_001009972.1 | 3, 4 | 770 | ||
| XM_006256125.2 | XP_006256187.1 | 3, 4 | 828 | |||
| GenBank mRNA | AY529104.1 | - | 3, 4 | 770 | ||
| BC105838.1 | - | 3, 4 | 778 | 7452960 | ||

Ggnbp1
Genomic Map