siRNA Details
| Gene Symbol: Naaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010967.2 | NP_001010967.1 | 3 | 509 | ||
| GenBank mRNA | AB162193.1 | - | 3 | 507 | ||
| BC105771.1 | - | 3 | 509 | 7930474 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Naaa| Gene Symbol: Naaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010967.2 | NP_001010967.1 | 3 | 509 | ||
| GenBank mRNA | AB162193.1 | - | 3 | 507 | ||
| BC105771.1 | - | 3 | 509 | 7930474 | ||