siRNA Details
| Gene Symbol: Eng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010968.2 | NP_001010968.1 | 3 | 638 | ||
| GenBank mRNA | AY562420.1 | - | 3 | 351 | ||
| BC105860.1 | - | 3 | 638 | 7456529 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eng| Gene Symbol: Eng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010968.2 | NP_001010968.1 | 3 | 638 | ||
| GenBank mRNA | AY562420.1 | - | 3 | 351 | ||
| BC105860.1 | - | 3 | 638 | 7456529 | ||