siRNA Details
| Gene Symbol: Xkr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012229.1 | NP_001012229.1 | 4 | 1500 | ||
| XM_006237733.2 | XP_006237795.1 | 4 | 1583 | |||
| GenBank mRNA | AY534263.1 | - | 4 | 1500 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xkr9| Gene Symbol: Xkr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012229.1 | NP_001012229.1 | 4 | 1500 | ||
| XM_006237733.2 | XP_006237795.1 | 4 | 1583 | |||
| GenBank mRNA | AY534263.1 | - | 4 | 1500 | ||