siRNA Details
| Gene Symbol: Rnase13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012231.1 | NP_001012231.1 | 1 | 340 | ||
| XM_006251896.2 | XP_006251958.1 | 2 | 2478 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnase13| Gene Symbol: Rnase13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012231.1 | NP_001012231.1 | 1 | 340 | ||
| XM_006251896.2 | XP_006251958.1 | 2 | 2478 | |||