siRNA Details
| Gene Symbol: Impact | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012235.1 | NP_001012235.1 | 8, 9 | 756 | ||
| GenBank mRNA | AY574213.1 | - | 8, 9 | 756 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Impact| Gene Symbol: Impact | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012235.1 | NP_001012235.1 | 8, 9 | 756 | ||
| GenBank mRNA | AY574213.1 | - | 8, 9 | 756 | ||