siRNA Details
| Gene Symbol: Fut10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012360.1 | NP_001012360.1 | 2, 3 | 385 | ||
| GenBank mRNA | AJ879584.1 | - | 2, 3 | 385 | ||
| AJ880010.1 | - | 2, 3 | 385 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fut10| Gene Symbol: Fut10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012360.1 | NP_001012360.1 | 2, 3 | 385 | ||
| GenBank mRNA | AJ879584.1 | - | 2, 3 | 385 | ||
| AJ880010.1 | - | 2, 3 | 385 | |||