siRNA Details
| Gene Symbol: Arid1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758744.1 | XP_008756966.1 | 3788 | |||
| GenBank mRNA | AJ005202.2 | - | 2468 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arid1b| Gene Symbol: Arid1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758744.1 | XP_008756966.1 | 3788 | |||
| GenBank mRNA | AJ005202.2 | - | 2468 | |||