siRNA Details
| Gene Symbol: Gucy1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017090.2 | NP_058786.2 | 8, 9 | 2004 | ||
| XM_006232518.2 | XP_006232580.1 | 9, 10 | 2596 | |||
| XM_008761107.1 | XP_008759329.1 | 7, 8 | 2627 | |||
| XM_008761108.1 | XP_008759330.1 | 8, 9 | 2370 | |||
| GenBank mRNA | AB096020.1 | - | 7, 8 | 1867 | ||
| BC085746.1 | - | 8, 9 | 2004 | 7190419 | ||
| M57405.1 | - | 8, 9 | 2323 | |||
| U60835.1 | - | 8, 9 | 2323 | |||

Gucy1a3
Genomic Map