siRNA Details
| Gene Symbol: Scn3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013119.1 | NP_037251.1 | 7 | 1365 | ||
| XM_008761914.1 | XP_008760136.1 | 6 | 1168 | |||
| XM_008761915.1 | XP_008760137.1 | 7 | 1171 | |||
| XM_008761916.1 | XP_008760138.1 | 6 | 1169 | |||
| XM_008761917.1 | XP_008760139.1 | 6 | 1168 | |||
| XM_008761918.1 | XP_008760140.1 | 6 | 1168 | |||
| XM_008761919.1 | XP_008760141.1 | 6 | 1168 | |||
| XM_008761920.1 | XP_008760142.1 | 6 | 1168 | |||
| GenBank mRNA | Y00766.1 | - | 7 | 1365 | ||

Scn3a
Genomic Map