siRNA Details
Gene Symbol: Rps7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC060557.1 | - | 6, 7 | 542 | 6888098 | |
BC168722.1 | - | 5, 6 | 522 | 6922220 | ||
X53377.1 | - | 6, 7 | 538 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rps7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC060557.1 | - | 6, 7 | 542 | 6888098 | |
BC168722.1 | - | 5, 6 | 522 | 6922220 | ||
X53377.1 | - | 6, 7 | 538 |