siRNA Details
| Gene Symbol: Cps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017072.1 | NP_058768.1 | 14 | 1420 | ||
| GenBank mRNA | M11710.1 | - | 14 | 1559 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cps1| Gene Symbol: Cps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017072.1 | NP_058768.1 | 14 | 1420 | ||
| GenBank mRNA | M11710.1 | - | 14 | 1559 | ||