siRNA Details
| Gene Symbol: C1qtnf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191918.1 | NP_001178847.1 | 4 | 946 | ||
| XM_006246141.2 | XP_006246203.1 | 4 | 1033 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1qtnf2| Gene Symbol: C1qtnf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191918.1 | NP_001178847.1 | 4 | 946 | ||
| XM_006246141.2 | XP_006246203.1 | 4 | 1033 | |||