siRNA Details
| Gene Symbol: Zfp62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246188.2 | XP_006246250.1 | 3 | 2341 | ||
| XM_006246189.2 | XP_006246251.1 | 2 | 2288 | |||
| XM_006246193.2 | XP_006246255.1 | 3 | 2337 | |||
| XM_008767726.1 | XP_008765948.1 | 2 | 5825 | |||
| XM_008767727.1 | XP_008765949.1 | 2 | 5821 | |||

Zfp62
Genomic Map