siRNA Details
| Gene Symbol: Uqcrq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025134.1 | NP_001020305.1 | 2 | 812 | ||
| GenBank mRNA | AY323237.1 | - | 2 | 812 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uqcrq| Gene Symbol: Uqcrq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025134.1 | NP_001020305.1 | 2 | 812 | ||
| GenBank mRNA | AY323237.1 | - | 2 | 812 | ||