siRNA Details
| Gene Symbol: Sowaha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_573089.5 | XP_573089.1 | 1 | 2234 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sowaha| Gene Symbol: Sowaha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_573089.5 | XP_573089.1 | 1 | 2234 | ||