siRNA Details
| Gene Symbol: Ccdc69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109031.1 | NP_001102501.1 | 6 | 567 | ||
| XM_006246393.2 | XP_006246455.1 | 7 | 673 | |||
| XM_008767703.1 | XP_008765925.1 | 6 | 897 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc69| Gene Symbol: Ccdc69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109031.1 | NP_001102501.1 | 6 | 567 | ||
| XM_006246393.2 | XP_006246455.1 | 7 | 673 | |||
| XM_008767703.1 | XP_008765925.1 | 6 | 897 | |||