siRNA Details
Gene Symbol: RGD1559732 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750838.3 | XP_003750886.1 | 4 | 593 | ||
XM_006246534.2 | XP_006246596.1 | 3 | 518 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1559732 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750838.3 | XP_003750886.1 | 4 | 593 | ||
XM_006246534.2 | XP_006246596.1 | 3 | 518 |