siRNA Details
| Gene Symbol: Mief2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246545.2 | XP_006246607.1 | 4 | 1631 | ||
| XM_008767888.1 | XP_008766110.1 | 3 | 3040 | |||
| XM_008767889.1 | XP_008766111.1 | 4 | 3689 | |||
| XM_573103.6 | XP_573103.3 | 4 | 2220 | |||

Mief2
Genomic Map