siRNA Details
| Gene Symbol: Slc47a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191920.1 | NP_001178849.1 | 16 | 1442 | ||
| XM_008767850.1 | XP_008766072.1 | 15 | 1459 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc47a2| Gene Symbol: Slc47a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191920.1 | NP_001178849.1 | 16 | 1442 | ||
| XM_008767850.1 | XP_008766072.1 | 15 | 1459 | |||