siRNA Details
| Gene Symbol: LARGE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004737.4 | NP_004728.1 | 7 | 1229 | ||
| NM_133642.3 | NP_598397.1 | 6 | 1166 | |||
| XM_005261831.3 | XP_005261888.1 | 7 | 1075 | |||
| XM_005261832.3 | XP_005261889.1 | 6 | 1030 | |||
| XM_011530510.2 | XP_011528812.1 | 7 | 1016 | |||
| XM_011530512.2 | XP_011528814.1 | 2 | 153 | |||
| XM_011530514.2 | XP_011528816.1 | 7 | 1277 | |||
| GenBank mRNA | AB011181.3 | - | 6 | 1081 | ||
| AJ007583.1 | - | 7 | 1139 | |||
| AK294766.1 | - | 6 | 852 | |||
| AK301808.1 | - | 2 | 144 | |||
| AK310462.1 | - | 2 | 153 | |||
| BC117425.1 | - | 5 | 718 | 40125976 | ||
| BC126404.1 | - | 5 | 718 | 8992120 | ||
| CR456510.1 | - | 5 | 678 | |||
| KJ534877.1 | - | 5 | 650 | |||
| KJ535033.1 | - | 5 | 650 | |||
| KJ535073.1 | - | 4 | 526 | |||
| KJ535074.1 | - | 4 | 567 | |||

LARGE1
Genomic Map