siRNA Details
Gene Symbol: Sco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173374.1 | NP_001166845.1 | 5, 6 | 762 | ||
GenBank mRNA | BC158882.1 | - | 5, 6 | 697 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
