siRNA Details
| Gene Symbol: Slc25a35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109033.2 | NP_001102503.1 | 4 | 1112 | ||
| XM_006246776.2 | XP_006246838.1 | 5 | 2011 | |||
| XM_008767852.1 | XP_008766074.1 | 3 | 2281 | |||
| GenBank mRNA | BC158839.1 | - | 4 | 1112 | 7387741 | |

Slc25a35
Genomic Map