siRNA Details
| Gene Symbol: LOC497938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017475.1 | NP_001017475.1 | 3 | 853 | ||
| XM_008767853.1 | XP_008766075.1 | 2 | 882 | |||
| GenBank mRNA | BC079298.1 | - | 3 | 853 | 7134515 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC497938| Gene Symbol: LOC497938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017475.1 | NP_001017475.1 | 3 | 853 | ||
| XM_008767853.1 | XP_008766075.1 | 2 | 882 | |||
| GenBank mRNA | BC079298.1 | - | 3 | 853 | 7134515 | |