siRNA Details
| Gene Symbol: Trpv3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025757.2 | NP_001020928.2 | 9 | 1249 | ||
| XM_008767859.1 | XP_008766081.1 | 9 | 3144 | |||
| XM_008767860.1 | XP_008766082.1 | 8 | 1283 | |||
| GenBank mRNA | AY325813.1 | - | 9 | 1268 | ||

Trpv3
Genomic Map