siRNA Details
| Gene Symbol: Tlcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246868.2 | XP_006246930.1 | 2 | 864 | ||
| XM_006246869.2 | XP_006246931.1 | 2 | 864 | |||
| XM_006246870.2 | XP_006246932.1 | 2 | 864 | |||
| XM_573142.6 | XP_573142.4 | 2 | 864 | |||
| GenBank mRNA | FQ219135.1 | - | 2 | 336 | ||

Tlcd2
Genomic Map