siRNA Details
| Gene Symbol: Fam222b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109038.1 | NP_001102508.1 | 3 | 273 | ||
| XM_006246944.1 | XP_006247006.1 | 3 | 292 | |||
| XM_008768094.1 | XP_008766316.1 | 2 | 770 | |||
| XM_008768095.1 | XP_008766317.1 | 3 | 903 | |||

Fam222b
Genomic Map