siRNA Details
| Gene Symbol: Lyrm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173471.1 | NP_001166942.1 | 2, 3 | 197 | ||
| GenBank mRNA | BC167116.1 | - | 2, 3 | 197 | 6922274 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lyrm9| Gene Symbol: Lyrm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173471.1 | NP_001166942.1 | 2, 3 | 197 | ||
| GenBank mRNA | BC167116.1 | - | 2, 3 | 197 | 6922274 | |