siRNA Details
| Gene Symbol: Myo19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163736.1 | NP_001157208.1 | 24, 25 | 2777 | ||
| XM_008768098.1 | XP_008766320.1 | 24, 25 | 2909 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myo19| Gene Symbol: Myo19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163736.1 | NP_001157208.1 | 24, 25 | 2777 | ||
| XM_008768098.1 | XP_008766320.1 | 24, 25 | 2909 | |||