siRNA Details
| Gene Symbol: Znhit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_573169.6 | XP_573169.3 | 3, 4 | 241 | ||
| GenBank mRNA | FQ231514.1 | - | 3, 4 | 218 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Znhit3| Gene Symbol: Znhit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_573169.6 | XP_573169.3 | 3, 4 | 241 | ||
| GenBank mRNA | FQ231514.1 | - | 3, 4 | 218 | ||