siRNA Details
| Gene Symbol: Fam117a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109039.1 | NP_001102509.1 | 8 | 1236 | ||
| XM_006247206.1 | XP_006247268.1 | 8 | 1231 | |||
| XM_006247207.2 | XP_006247269.1 | 7 | 1097 | |||
| GenBank mRNA | BC168983.1 | - | 6 | 1301 | 7387439 | |

Fam117a
Genomic Map