siRNA Details
| Gene Symbol: Zfp652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080207.1 | NP_001073676.1 | 1 | 856 | ||
| GenBank mRNA | BC129097.1 | - | 1 | 856 | 8365300 | |
| Gene Symbol: LOC685279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763583.1 | XP_008761805.1 | 1 | 428 | ||

Zfp652
Genomic Map