siRNA Details
| Gene Symbol: Hoxb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191925.1 | NP_001178854.1 | 1 | 179 | ||
| XM_008768104.1 | XP_008766326.1 | 1 | 2012 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008268, BC103604, BC103607, BC103595, BC103596, and others.

Hoxb5
Genomic Map